accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNNNNNNOCH3OH3COH3COCH3OCH3ClCl

CAS RN 106629-90-7

IUPAC Name: 5-[(3,4-dimethoxyphenyl)methyl]-2-(4'-{5-[(3,4-dimethoxyphenyl)methyl]-3-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl}-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl)-3-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Synonyms: 5-[(3,4-dimethoxyphenyl)methyl]-2-(4'-{5-[(3,4-dimethoxyphenyl)methyl]-3-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl}-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl)-3-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular Weight (g/mol): 875.81
Molecular Formula: C46H44Cl2N8O6
InChi Key: WYENYHSISIKTHQ-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].COC1=C(OC)C=C(CC2=NN(C3=CC=CC=C3)[N+](=N2)C2=C(OC)C=C(C=C2)C2=CC(OC)=C(C=C2)[N+]2=NC(CC3=CC(OC)=C(OC)C=C3)=NN2C2=CC=CC=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results