CAS RN 107-15-3
Ethylenediamine, 99+%
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine Anhydrous 98.0+%, TCI America™
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, 99%
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, 99+%, AcroSeal™
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, 99%
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, 70%, Technical Grade, Ricca Chemical
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): Mixture MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Ethylenediamine, Technical Grade, 70% (w/v) ±3% (w/v), LabChem™
CAS: 107-15-3 Molecular Formula: C2H8N2 Molecular Weight (g/mol): 60.10 MDL Number: MFCD00008204 InChI Key: PIICEJLVQHRZGT-UHFFFAOYSA-N Synonym: ethylenediamine,1,2-ethanediamine,1,2-diaminoethane,ethylene diamine,ethylenediamine,dimethylenediamine,edamine,1,2-ethylenediamine,aethaldiamin,aethylenediamin PubChem CID: 3301 ChEBI: CHEBI:30347 IUPAC Name: ethane-1,2-diamine SMILES: NCCN
Sigma Aldrich Ethylenediamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethylenediamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2-Aminoethyl Diphenylborinate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethylenediamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Biphenyl Sodium Complex Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethyl Diazoacetate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2