Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine, Technical Grade, 70% (w/v) ±3% (w/v), LabChem™

Catalog No. LC141402 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC141402 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC141402 Supplier LabChem, Inc. Supplier No. LC141402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-15-3 , 7732-18-5
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

CAS Max % 70, 30
Color Colorless to Yellow
Odor Ammonia
Packaging Poly Bottle
Linear Formula C2H8N2
Quantity 1 L
UN Number UN1604
Solubility Information Soluble in water
Formula Weight 60.1
Percent Purity 70% ±3% (w/v)
Grade Technical
Physical Form Liquid
Chemical Name or Material Ethylenediamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.