accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107220-28-0

IUPAC Name: hydrogen (2R,5'R)-5'-methyl-4-azaspiro[bicyclo[2.2.2]octane-2,2'-[1,4]oxathiolane] chloride
Synonyms: hydrogen cevimeline chloride
Molecular Weight (g/mol): 235.77
Molecular Formula: C10H18ClNOS
InChi Key: SURWTGAXEIEOGY-GHXDPTCOSA-N
SMILES: [H+].[Cl-].C[C@@H]1O[C@@]2(CS1)CN1CCC2CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results