accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNO

CAS RN 107259-06-3

IUPAC Name: tert-butyl N-(1-formylcyclopropyl)carbamate
Synonyms: tert-butyl N-(1-formylcyclopropyl)carbamate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: ACMHCEYCNRURST-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1(CC1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results