accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NFH3C

CAS RN 107264-00-6

IUPAC Name: 1-fluoro-2,4,6-trimethylpyridin-1-ium
Synonyms: 1-fluoro-2,4,6-trimethylpyridin-1-ium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: JMEOVHKKRMLNBM-UHFFFAOYSA-N
SMILES: CC1=CC(C)=[N+](F)C(C)=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)