Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fluoro-2,4,6-trimethylpyridinium Triflate 85-90%, Thermo Scientific™

Catalog No. AC259915000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259915000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259915000 Supplier Thermo Scientific Chemicals Supplier No. 259915000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107264-00-6
Molecular Formula C9H11F4NO3S
Molecular Weight (g/mol) 289.245
InChI Key PRIGFEJKMMRJSF-UHFFFAOYSA-M
Synonym 1-fluoro-2,4,6-trimethylpyridinium triflate, n-fluoro-2,4,6-trimethylpyridinium triflate, 1-fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium triflate, 1-fluoro-2,4,6-trimethylpyridinium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium; trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridine, trifluoromethanesulfonic acid, acmc-1bprl, 1-fluoro-2,4,6-trimethylpyridiniumtriflate, 1-fluoro-2,4,6-trimethyl-pyridinium triflate
PubChem CID 2724575
IUPAC Name 1-fluoro-2,4,6-trimethylpyridin-1-ium;trifluoromethanesulfonate
SMILES CC1=CC(=[N+](C(=C1)C)F)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

CAS Min % 85.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Quantity 500 mg
Formula Weight 289.23
Percent Purity ≥85%
Physical Form Crystalline Powder
Chemical Name or Material N-Fluoro-2, 4, 6-trimethylpyridinium triflate, 85-90%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.