accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073371-87-5

IUPAC Name: 2-propoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-propoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 263.14
Molecular Formula: C14H22BNO3
InChi Key: NEKFTDDYHKYVSI-UHFFFAOYSA-N
SMILES: CCCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results