Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propoxypyridine-3-boronic acid pinacol ester, Thermo Scientific™

Catalog No. AAH5010906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5010906 5 g
AAH5010903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5010906 Supplier Thermo Scientific Chemicals Supplier No. H5010906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-87-5
Molecular Formula C14H22BNO3
Molecular Weight (g/mol) 263.144
MDL Number MFCD07781169
InChI Key NEKFTDDYHKYVSI-UHFFFAOYSA-N
Synonym 2-propoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-n-propoxypyridine-3-boronic acid pinacol ester, 2-propoxypyridine-3-boronic acid pinacol ester, 2-propoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-propoxypyridine-3-boronic acid, pinacol ester, 2-n-propoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 24208791
IUPAC Name 2-propoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)OCCC

Specifications

Quantity 5 g
Formula Weight 263.15
Chemical Name or Material 2-n-Propoxypyridine-3-boronic acid pinacol ester
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.