accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOH

CAS RN 107430-66-0

IUPAC Name: 4-[7-(adamantan-1-yl)-6-hydroxynaphthalen-2-yl]benzoic acid
Synonyms: 4-[7-(adamantan-1-yl)-6-hydroxynaphthalen-2-yl]benzoic acid
Molecular Weight (g/mol): 398.5
Molecular Formula: C27H26O3
InChi Key: VCQGNUWOMLYNNG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC2=CC(=C(O)C=C2C=C1)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)