accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107703-78-6

HON

CAS RN 107703-78-6

IUPAC Name: phenyl[1-(2-phenylethyl)piperidin-4-yl]methanol
Synonyms: glemanserin 格来色林
Molecular Weight (g/mol): 295.43
Molecular Formula: C20H25NO
InChi Key: AXNGJCOYCMDPQG-UHFFFAOYNA-N
SMILES: OC(C1CCN(CCC2=CC=CC=C2)CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)