accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3HO

CAS RN 108-01-0

IUPAC Name: 2-(dimethylamino)ethan-1-ol
Synonyms: dimethylaminoethanol DMAE DMEA deanol (substance) norcholine amietol M 21 n,n dimethyl 2 hydroxyethylamine astyl deanol deanol acetamidobenzoate
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: UEEJHVSXFDXPFK-UHFFFAOYSA-N
SMILES: CN(C)CCO
Molecular Weight (g/mol) 
  • (5)
  • (16)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (4)
  • (4)
Boiling Point 
  • (4)
  • (2)
  • (2)
  • (8)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (1)
  • (8)
  • (4)
  • (1)
  • (2)
  • (6)
  • (5)
  • (16)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (8)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (10)
  • (1)
  • (4)
  • (1)