Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylethanolamine, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611867 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
200 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-867 500 mL Glass Bottle
18-611-864 2.5 L Glass Bottle
18-611-865 20 L Glass Bottle
18-611-866 200 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-867 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2399500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
IUPAC Name 2-(dimethylamino)ethan-1-ol
SMILES CN(C)CCO

Specifications

CAS Max % 1
Boiling Point 133°C to 139°C
Packaging Glass Bottle
Quantity 500 mL
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material N,N-Dimethylethanolamine

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.