Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)ethanol, TCI America™
SDP

Catalog No. D0649500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0649500ML 500 mL
D064925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0649500ML Supplier TCI America Supplier No. D0649500ML

Chemical Identifiers

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002846
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon
PubChem CID 7902
ChEBI CHEBI:271436
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Specifications

Color Yellow
Boiling Point 134°C
Quantity 500 mL
UN Number 2051
Formula Weight 89.14
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethanol
EINECSNumber (2)-0297&(2)-0353
RTECSNumber KK6125000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.