accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-18-9

IUPAC Name: bis(propan-2-yl)amine
Synonyms: diisopropylamine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: UAOMVDZJSHZZME-UHFFFAOYSA-N
SMILES: CC(C)NC(C)C
Boiling Point 
  • (2)
  • (11)
  • (5)
  • (5)
Molecular Weight (g/mol) 
  • (9)
  • (11)
  • (3)
Quantity 
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (5)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (2)
  • (11)
  • (5)
  • (7)
  • (2)
  • (11)
  • (5)
  • (5)
  • (9)
  • (11)
  • (3)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (10)
  • (5)
  • (2)