accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COClO

CAS RN 108-23-6

IUPAC Name: propan-2-yl carbonochloridate
Synonyms: isopropyl chloroformate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: IVRIRQXJSNCSPQ-UHFFFAOYSA-N
SMILES: CC(C)OC(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)