Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Chloroformate (ca. 30% in Toluene, ca. 2mol/L), TCI America™
SDP

Catalog No. I0768500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0768500G 500 g
I0768100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0768500G Supplier TCI America Supplier No. I0768500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-23-6
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00075068
InChI Key IVRIRQXJSNCSPQ-UHFFFAOYSA-N
Synonym isopropyl chloroformate, isopropyl chlorocarbonate, carbonochloridic acid, 1-methylethyl ester, isopropyl chloromethanoate, isopropyl carbonochloridate, chloroformic acid isopropyl ester, unii-8lg1g18vxr, formic acid, chloro-, isopropyl ester, carbonochloride acid, 1-methylethyl ester, isopropylester kyseliny chlormravenci
PubChem CID 7917
IUPAC Name propan-2-yl carbonochloridate
SMILES CC(C)OC(=O)Cl

Specifications

Color Colorless
Quantity 500 g
UN Number 3286
Formula Weight 122.55
Physical Form Liquid
Chemical Name or Material Isopropyl Chloroformate (ca. 30% in Toluene, ca. 2mol/L)
EINECSNumber (2)-1144&(2)-1704
RTECSNumber LQ6475000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.