CAS RN 108-38-3
m-Xylene, 99+%, extra pure
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene, 99%
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
M-Xylene, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.
m-Xylene, 99%, Extra Dry, AcroSeal™
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene, 98%, Spectrum™ Chemical
CAS: 108-38-3 Molecular Weight (g/mol): 106.17
m-Xylene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™
m-Xylene is an aromatic liquid, which is an isomeric form of xylene.
m-Xylene, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. m-Xylene is widely used as a residual solvent (RS) or organic volatile impurity (OVI) in pharmaceutical formulations.
m-Xylene 99.0+%, TCI America™
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene, SPEX CertiPrep™
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene (Synthesis Grade), Thermo Scientific™
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene, EMD-Millipore
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: 1, 3-Dimethylbenzene PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene, ≥99% (GC), Solstice
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N Synonym: m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150 PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1
m-Xylene in Dimethyl Sulfoxide, SPEX CertiPrep™
CAS: 108-38-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008536 InChI Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N PubChem CID: 7929 ChEBI: CHEBI:28488 IUPAC Name: 1,3-xylene SMILES: CC1=CC(C)=CC=C1