Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene, 98%, Spectrum™ Chemical
SDP

Catalog No. 18611837 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-837 500 mL Amber Glass Bottle
18-611-836 4 L Amber Glass Bottle
18-611-834 20 L Steel Can
18-611-835 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-837 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. X1015500MLGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-38-3
Molecular Weight (g/mol) 106.17

Specifications

Quantity 500 mL
Packaging Amber Glass Bottle
Boiling Point 137°C to 139°C
Percent Purity 0.98
Grade Reagent
Chemical Name or Material m-Xylene

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.