Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene, ≥99% (GC), Solstice

Catalog No. 60048502 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-502 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-502 Supplier Solstice Supplier No. 956721L

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.0% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -48°C
Density 0.868g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Ignition Point 982°F
Packaging Glass Bottle
Refractive Index 1.497
Linear Formula C6H4(CH3)2
UN Number UN1307
Beilstein 605441
Boiling Point 138°C to 139°C
Formula Weight 106.17g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material m-Xylene
Show More Show Less
Hazard Statement P261-P273-P280-P301 + P310-P305 + P351 + P338-P331
DOTInformation class 3 - PG 3 - Xylenes
RTECSNumber 203-576-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.