CAS RN 108-39-4
m-Cresol, 99%
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002302 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N Synonym: 3-Methylphenol PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol
m-Cresol, 99%
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002302 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N Synonym: m-cresol,metacresol,meta-cresol,phenol, 3-methyl,3-hydroxytoluene,m-methylphenol,3-cresol,m-kresol,m-oxytoluene,m-toluol PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol SMILES: CC1=CC(=CC=C1)O
m-Cresol 98.0+%, TCI America™
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002302 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N Synonym: m-cresol,metacresol,meta-cresol,phenol, 3-methyl,3-hydroxytoluene,m-methylphenol,3-cresol,m-kresol,m-oxytoluene,m-toluol PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol SMILES: CC1=CC(=CC=C1)O
m-Cresol, ≥99% (GC), MilliporeSigma™ Supelco™
m-Cresol is an isomer form of Cresol and is mainly used as an intermediate in the manufacture of chemicals. It is toxic in nature. It is a component of coal tar creosote which is used for wood preservation. It also possess bactericide and fungicide property.
m-Cresol, High Purity, 99%, Spectrum™ Chemical
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N IUPAC Name: 3-methylphenol SMILES: CC1=CC=CC(O)=C1
m-Cresol, MilliporeSigma™
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N Synonym: m-cresol,metacresol,meta-cresol,phenol, 3-methyl,3-hydroxytoluene,m-methylphenol,3-cresol,m-kresol,m-oxytoluene,m-toluol PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol SMILES: CC1=CC(=CC=C1)O
3-Methylphenol, SPEX CertiPrep™
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol SMILES: CC1=CC(=CC=C1)O
Metacresol, EP, USP, 98-102%, Spectrum™ Chemical
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N IUPAC Name: 3-methylphenol SMILES: CC1=CC=CC(O)=C1