Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol, High Purity, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604242 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-242 200 L Drum
18-604-238 100 g Amber Glass Bottle
18-604-239 500 g Amber Glass Bottle
18-604-241 20 L Poly Drum
18-604-240 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-242 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1367200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C1367, 108-39-4, C7H8O

Spectrum™ Chemical m-Cresol, High Purity is a colorless organic compound and liquid. It is derived from phenol and is commonly extracted from coal tar. It is also known as 3-methylphenol. High purity products supplied by Spectrum are indicative of a grade suitable for research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
IUPAC Name 3-methylphenol
SMILES CC1=CC=CC(O)=C1

Specifications

CAS Max % 100%
Quantity 200 L
Packaging Drum
UN Number UN2076
Percent Purity 99%
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.