Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metacresol, EP, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604338 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-338 100 g Amber Glass Bottle
18-604-337 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-604-338 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C2773100GM

missing translation for 'validMainframeMsg'

C2773, 108-39-4, C7H8O

Spectrum™ Chemical Metacresol, USP, EP is a local antiseptic with a high germicidal power. All Spectrum Chemical USP, EP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
IUPAC Name 3-methylphenol
SMILES CC1=CC=CC(O)=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2076
Percent Purity 98 to 102%
Grade Multi Compendia
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.