accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1080-32-6

IUPAC Name: diethyl benzylphosphonate
Synonyms: diethyl benzylphosphonate
Molecular Weight (g/mol): 228.23
Molecular Formula: C11H17O3P
InChi Key: AIPRAPZUGUTQKX-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=CC=C1)OCC
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (4)