accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)HO(S)O(R)H3CNHNON

CAS RN 108068-98-0

IUPAC Name: hexyl (15R,16S,18S)-16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Synonyms: hexyl (15R,16S,18S)-16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Molecular Weight (g/mol): 537.62
Molecular Formula: C32H31N3O5
InChi Key: ZHEHVZXPFVXKEY-RUAOOFDTSA-N
SMILES: CCCCCCOC(=O)[C@]1(O)C[C@@H]2O[C@@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
Form 
  • (1)
  • (2)
Product Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results