accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108923-79-1

IUPAC Name: bis[amino(methylsulfanyl)methylidene]butanedinitrile
Synonyms: bis[amino(methylsulfanyl)methylidene]butanedinitrile
Molecular Weight (g/mol): 226.32
Molecular Formula: C8H10N4S2
InChi Key: LBQNBMSPTURKGS-UHFFFAOYSA-N
SMILES: CSC(N)=C(C#N)C(C#N)=C(N)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results