Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U0124, Tocris Bioscience™
SDP

Catalog No. 186810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-681-0 Supplier R&D Systems Supplier No. 1868/10
This item is not returnable. View return policy

Inactive analog of U0126 (Cat. No. 1144)

Inactive analog of U0126 (Cat. No. 1144). Does not inhibit MEK at concentrations up to 100 μM. Used as a negative control.

Active Analog also available.

Chemical Identifiers

CAS 108923-79-1
Molecular Formula C8H10N4S2
Molecular Weight (g/mol) 226.316
InChI Key LBQNBMSPTURKGS-KQQUZDAGSA-N
Synonym 1,4-diamino-1,4-bismethylthio-butadien-2,3-dicarbonitril, chembl97858, bis amino methylthio methylene butanedinitrile, 2z,3z-2,3-bis amino methylsulfanyl methylidene butanedinitrile
PubChem CID 5842955
IUPAC Name (2Z,3Z)-2,3-bis[amino(methylsulfanyl)methylidene]butanedinitrile
SMILES CSC(=C(C#N)C(=C(N)SC)C#N)N

Specifications

Quantity 10 mg
Formula Weight 226.31
Percent Purity >98%
Chemical Name or Material U0124
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.