accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108930-17-2

IUPAC Name: dihydrogen 5-[(2-methylpiperazin-1-yl)sulfonyl]isoquinoline dichloride
Synonyms: dihydrogen 5-(2-methylpiperazin-1-ylsulfonyl)isoquinoline dichloride
Molecular Weight (g/mol): 364.29
Molecular Formula: C14H19Cl2N3O2S
InChi Key: OARGPFMFRLLKPF-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1CNCCN1S(=O)(=O)C1=C2C=CN=CC2=CC=C1
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results