accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOOSOOCH3OSOOCH3

CAS RN 108963-16-2

IUPAC Name: 1,4-bis(methanesulfonyloxy)butan-2-yl methanesulfonate
Synonyms: 1,4-bis(methanesulfonyloxy)butan-2-yl methanesulfonate
Molecular Weight (g/mol): 340.38
Molecular Formula: C7H16O9S3
InChi Key: YJIUOKPKBPEIMF-UHFFFAOYNA-N
SMILES: CS(=O)(=O)OCCC(COS(C)(=O)=O)OS(C)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results