Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Tris(methanesulfonyloxy)butane, TCI America™
SDP

Catalog No. T145425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T145425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T145425G Supplier TCI America Supplier No. T145425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108963-16-2
Molecular Formula C7H16O9S3
Molecular Weight (g/mol) 340.376
MDL Number MFCD00191688
InChI Key YJIUOKPKBPEIMF-UHFFFAOYSA-N
Synonym 1,2,4-Butanetriol Trimethanesulfonate
PubChem CID 13504422
IUPAC Name 3,4-bis(methylsulfonyloxy)butyl methanesulfonate
SMILES CS(=O)(=O)OCCC(COS(=O)(=O)C)OS(=O)(=O)C

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 25 g
Formula Weight 340.38
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Tris(methanesulfonyloxy)butane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.