CAS RN 109-99-9
Tetrahydrofuran Non UV, MilliporeSigma™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, anhydrous, 99.9%, ACS Grade, DriSolv, MilliporeSigma™
Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran (HPLC), Fisher Chemical™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.9% min., MilliporeSigma™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, anhydrous, 99.8+%, unstab.
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran Anhydrous (stabilized with BHT) 99.5+%, TCI America™
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1
Tetrahydrofuran, 99.9%, for HPLC, unstabilized, Thermo Scientific Chemicals
CAS: 109-99-9 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00005356 InChI Key: WYURNTSHIVDZCO-UHFFFAOYSA-N Synonym: tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano PubChem CID: 8028 ChEBI: CHEBI:26911 IUPAC Name: oxolane SMILES: C1CCOC1