CAS RN 109089-77-2
IUPAC Name:
3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid
Synonyms:
3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid
Molecular Weight (g/mol):
270.33
Molecular Formula:
C17H18O3
InChi Key:
KSLPLDLDQANKOD-UHFFFAOYNA-N
SMILES:
COC1=C(C=C(C)C=C1)C(CC(O)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3-(2-Methoxy-5-methylphenyl)-3-phenylpropionic Acid 98.0+%, TCI America™
CAS: 109089-77-2 Molecular Formula: C17H18O3 Molecular Weight (g/mol): 270.328 MDL Number: MFCD01098026 InChI Key: KSLPLDLDQANKOD-UHFFFAOYSA-N PubChem CID: 3468322 IUPAC Name: 3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid SMILES: CC1=CC(=C(C=C1)OC)C(CC(=O)O)C2=CC=CC=C2