accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONNNOSOONNCl

CAS RN 1092351-10-4

IUPAC Name: 8-(4-{[4-(4-chlorophenyl)piperazin-1-yl]sulfonyl}phenyl)-1-propyl-2,6-dihydro-1H-purine-2,6-dione
Synonyms: 8-{4-[4-(4-chlorophenyl)piperazin-1-ylsulfonyl]phenyl}-1-propylpurine-2,6-dione
Molecular Weight (g/mol): 527
Molecular Formula: C24H23ClN6O4S
InChi Key: OHVMMGQPJADDAJ-UHFFFAOYSA-N
SMILES: CCCN1C(=O)N=C2N=C(N=C2C1=O)C1=CC=C(C=C1)S(=O)(=O)N1CCN(CC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)