accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109531-96-6

IUPAC Name: N-hydroxy-N-methylfuran-2-carboxamide
Synonyms: N-hydroxy-N-methylfuran-2-carboxamide
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: KCAHIEOWHDMBKM-UHFFFAOYSA-N
SMILES: CN(O)C(=O)C1=CC=CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results