CAS RN 110-19-0
Isobutyl acetate, 98%
CAS: 110-19-0 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00008932 InChI Key: GJRQTCIYDGXPES-UHFFFAOYSA-N Synonym: isobutyl acetate,isobutyl ethanoate,acetic acid, 2-methylpropyl ester,2-methyl-1-propyl acetate,2-methylpropyl ethanoate,acetic acid, isobutyl ester,acetic acid isobutyl ester,acetate d'isobutyle,iso-butyl acetate,beta-methylpropyl ethanoate PubChem CID: 8038 ChEBI: CHEBI:50569 IUPAC Name: 2-methylpropyl acetate SMILES: CC(C)COC(C)=O
Isobutyl Acetate 99.0+%, TCI America™
CAS: 110-19-0 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00008932 InChI Key: GJRQTCIYDGXPES-UHFFFAOYSA-N Synonym: isobutyl acetate,isobutyl ethanoate,acetic acid, 2-methylpropyl ester,2-methyl-1-propyl acetate,2-methylpropyl ethanoate,acetic acid, isobutyl ester,acetic acid isobutyl ester,acetate d'isobutyle,iso-butyl acetate,beta-methylpropyl ethanoate PubChem CID: 8038 ChEBI: CHEBI:50569 IUPAC Name: 2-methylpropyl acetate SMILES: CC(C)COC(C)=O
Isobutyl acetate, 98%
CAS: 110-19-0 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00008932 InChI Key: GJRQTCIYDGXPES-UHFFFAOYSA-N Synonym: isobutyl acetate,isobutyl ethanoate,acetic acid, 2-methylpropyl ester,2-methyl-1-propyl acetate,2-methylpropyl ethanoate,acetic acid, isobutyl ester,acetic acid isobutyl ester,acetate d'isobutyle,iso-butyl acetate,beta-methylpropyl ethanoate PubChem CID: 8038 ChEBI: CHEBI:50569 IUPAC Name: 2-methylpropyl acetate SMILES: CC(C)COC(C)=O
Isobutyl acetate, Analytical Standard, >99.8% (GC), MilliporeSigma™ Supelco™
Isobutyl acetate is a chemical compound, commonly used as a solvent and flavoring agent in food products.
Isobutyl Acetate, SPEX CertiPrep™
CAS: 110-19-0 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00008932 InChI Key: GJRQTCIYDGXPES-UHFFFAOYSA-N PubChem CID: 8038 ChEBI: CHEBI:50569 IUPAC Name: 2-methylpropyl acetate SMILES: CC(C)COC(C)=O
MP Biomedicals, Inc Isobutyl Acetate, MP Biomedicals
CAS: 110-19-0 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00008932 InChI Key: GJRQTCIYDGXPES-UHFFFAOYSA-N Synonym: isobutyl acetate,isobutyl ethanoate,acetic acid, 2-methylpropyl ester,2-methyl-1-propyl acetate,2-methylpropyl ethanoate,acetic acid, isobutyl ester,acetic acid isobutyl ester,acetate d'isobutyle,iso-butyl acetate,beta-methylpropyl ethanoate PubChem CID: 8038 ChEBI: CHEBI:50569 IUPAC Name: 2-methylpropyl acetate SMILES: CC(C)COC(C)=O