CAS RN 110-76-9
IUPAC Name:
2-ethoxyethan-1-aminium
Synonyms:
2-ethoxyethanaminium
Molecular Weight (g/mol):
90.15
Molecular Formula:
C4H12NO
InChi Key:
BPGIOCZAQDIBPI-UHFFFAOYSA-O
SMILES:
CCOCC[NH3+]
1
–
1
of
1
results
1
2-Ethoxyethylamine 98.0+%, TCI America™
CAS: 110-76-9 Molecular Formula: C4H11NO Molecular Weight (g/mol): 89.138 MDL Number: MFCD00025604 InChI Key: BPGIOCZAQDIBPI-UHFFFAOYSA-N Synonym: 2-Aminoethyl Ethyl Ether, O-Ethylethanolamine PubChem CID: 66970 IUPAC Name: 2-ethoxyethanamine SMILES: CCOCCN