Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethylamine 98.0+%, TCI America™
SDP

Catalog No. E022325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E022325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E022325ML Supplier TCI America Supplier No. E022325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-76-9
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00025604
InChI Key BPGIOCZAQDIBPI-UHFFFAOYSA-N
Synonym 2-Aminoethyl Ethyl Ether, O-Ethylethanolamine
PubChem CID 66970
IUPAC Name 2-ethoxyethanamine
SMILES CCOCCN

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 mL
UN Number 2733
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.