CAS RN 110-91-8
Morpholine, 99%
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, 99+%, ACS reagent
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, 99+%, extra pure
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, ACS, 99.0% min
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, ≥99.5% (GC), MilliporeSigma™ Supelco™
Morpholines are systemic compounds that exert both protective and curative actions against fungal diseases.
Morpholine, ACS, 99%, Spectrum™ Chemical
CAS: 110-91-8 Molecular Weight (g/mol): 87.12 g/mol
Morpholine, ACS Reagent Grade, Ricca Chemical
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine Standard, 1000 ppm C4H9NO, Ricca Chemical
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine, ACS Grade, ≥ 99.0 %, LabChem™
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Morpholine Standard, Certified, 1000ppm ±10ppm (1mL = 1mg), LabChem™
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
Sigma Aldrich Morpholine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Morpholine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Morpholine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Morpholine (Certified ACS), Fisher Chemical
CAS: 110-91-8 Molecular Formula: C4H9NO Molecular Weight (g/mol): 87.12 MDL Number: MFCD00005972 InChI Key: YNAVUWVOSKDBBP-UHFFFAOYSA-N Synonym: 1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro PubChem CID: 8083 ChEBI: CHEBI:34856 IUPAC Name: morpholine SMILES: C1COCCN1
- 1
- 2