Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine (Certified ACS), Fisher Chemical

Catalog No. M2631 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M263-1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M263-1

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Boiling Range 4.0°C max.
Melting Point -5°C
Color Colorless
Density 0.990g/cm³
Boiling Point 126°C
Flash Point 32°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Linear Formula NH(CH2)2OCH2CH2
Quantity 1 L
UN Number 2054
Vapor Density 3.0g/mol
Viscosity 2.23 cP at 20°C
Vapor Pressure 11mbar at 20°C
Formula Weight 87.12
Percent Purity ≥99%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Hygroscopic. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Do not breathe vapors/dust. Do not ingest. Take precautionary measures against static discharges. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:1
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.