accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOOHOONOO

CAS RN 1101854-58-3

IUPAC Name: 4-nitrophenyl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Synonyms: 4-nitrophenyl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Molecular Weight (g/mol): 520.49
Molecular Formula: C27H24N2O9
InChi Key: SEGYOKHGGFKMCX-UHFFFAOYSA-N
SMILES: OC(C1CCN(CC1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O)(C1=CC2=C(OCO2)C=C1)C1=CC2=C(OCO2)C=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results