CAS RN 110283-66-4
IUPAC Name:
hydrogen 1-(2-{bis[4-(trifluoromethyl)phenyl]methoxy}ethyl)-1,2,5,6-tetrahydropyridine-3-carboxylic acid chloride
Synonyms:
hydrogen 1-(2-{bis[4-(trifluoromethyl)phenyl]methoxy}ethyl)-5,6-dihydro-2H-pyridine-3-carboxylic acid chloride
Molecular Weight (g/mol):
509.87
Molecular Formula:
C23H22ClF6NO3
InChi Key:
NUQWSOWKRTZJTO-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].OC(=O)C1=CCCN(CCOC(C2=CC=C(C=C2)C(F)(F)F)C2=CC=C(C=C2)C(F)(F)F)C1
1
–
1
of
1
results
1
R&D Systems™ CI 966 hydrochloride
CAS: 110283-66-4 Molecular Formula: C23H22ClF6NO3