accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(S)(R)HNPOHOHO

CAS RN 110347-85-8

IUPAC Name: (2S,4R)-4-(phosphonomethyl)piperidine-2-carboxylic acid
Synonyms: selfotel
Molecular Weight (g/mol): 223.17
Molecular Formula: C7H14NO5P
InChi Key: LPMRCCNDNGONCD-RITPCOANSA-N
SMILES: OC(=O)[C@@H]1C[C@H](CP(O)(O)=O)CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results