accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNPOOHHONaNa

CAS RN 1103672-43-0

IUPAC Name: [(4-tetradecanamidophenyl)methyl]phosphonic acid disodium
Synonyms: (4-tetradecanamidophenyl)methylphosphonic acid disodium
Molecular Weight (g/mol): 443.48
Molecular Formula: C21H36NNa2O4P
InChi Key: LPESGHKFBOKCSG-UHFFFAOYSA-N
SMILES: [Na].[Na].CCCCCCCCCCCCCC(=O)NC1=CC=C(CP(O)(O)=O)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results