accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3CH3CCH3OOHO(E)

CAS RN 110368-33-7

IUPAC Name: 4-[(1E)-3-(3,5-di-tert-butylphenyl)-3-oxoprop-1-en-1-yl]benzoic acid
Synonyms: 4-[(1E)-3-(3,5-di-tert-butylphenyl)-3-oxoprop-1-en-1-yl]benzoic acid
Molecular Weight (g/mol): 364.49
Molecular Formula: C24H28O3
InChi Key: FOUVTBKPJRMLPE-FMIVXFBMSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1)C(=O)\C=C\C1=CC=C(C=C1)C(O)=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)