accessibility menu, dialog, popup

UserName

Viewing profile as user:
ISOONHN

CAS RN 110448-33-4

IUPAC Name: 1-[(5-iodonaphthalen-1-yl)sulfonyl]-1,4-diazepane
Synonyms: 1-(5-iodonaphthalen-1-ylsulfonyl)-1,4-diazepane
Molecular Weight (g/mol): 416.28
Molecular Formula: C15H17IN2O2S
InChi Key: GEHJIACZUFWBTK-UHFFFAOYSA-N
SMILES: IC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)N1CCCNCC1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results