CAS RN 1109276-89-2
CAS RN 1109276-89-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PF 04620110
Potent and selective DGAT-1 inhibitor; orally bioavailable