accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-21-7

IUPAC Name: 2-{2-[2-(acetyloxy)ethoxy]ethoxy}ethyl acetate
Synonyms: ethylenebis(oxyethylene)diacetate
Molecular Weight (g/mol): 234.25
Molecular Formula: C10H18O6
InChi Key: OVOUKWFJRHALDD-UHFFFAOYSA-N
SMILES: CC(=O)OCCOCCOCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results