Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Diacetate 98.0+%, TCI America™
SDP

Catalog No. T0923500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0923500G 500 g
T092325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0923500G Supplier TCI America Supplier No. T0923500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-21-7
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00026213
InChI Key OVOUKWFJRHALDD-UHFFFAOYSA-N
Synonym triethylene glycol diacetate, tdac, triglycol diacetate, triethylene glycol, diacetate, triglycol, diacetate, acetic acid, triethylene glycol diester, triethyleneglycoldiacetate, 2,2'-ethylenedioxy di ethyl acetate, ethanol, 2,2'-ethylenedioxydi-, diacetate, ethanol, 2,2'-1,2-ethanediylbis oxy bis-, diacetate
PubChem CID 8098
IUPAC Name 2-[2-(2-acetyloxyethoxy)ethoxy]ethyl acetate
SMILES CC(=O)OCCOCCOCCOC(=O)C

Specifications

Color Yellow
Boiling Point 300°C
Quantity 500 g
Formula Weight 234.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Diacetate
EINECSNumber (2)-2540
RTECSNumber YE4950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.