accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 111-44-4

IUPAC Name: 1-chloro-2-(2-chloroethoxy)ethane
Synonyms: BCEE 2,2'-dicloroetiletere caswell no. 309 2,2'-dichlorodiethyl ether epa pesticide chemical code 029501 2,2'-dichlor-diaethylaether bis(chloro-2-ethyl) oxide clorex β,β-dichlorodiethyl ether ccris 88
Molecular Weight (g/mol): 143.01
Molecular Formula: C4H8Cl2O
InChi Key: ZNSMNVMLTJELDZ-UHFFFAOYSA-N
SMILES: ClCCOCCCl
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)