CAS RN 111-90-0
Diethylene glycol monoethyl ether, 98%
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 MDL Number: MFCD00002872 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: diethylene glycol monoethyl ether,2-2-ethoxyethoxy ethanol,carbitol,transcutol,ethoxy diglycol,dioxitol,ethyl carbitol,ethyl digol,carbitol solvent,ethanol, 2-2-ethoxyethoxy PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™ Chemical
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.18 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N IUPAC Name: 2-(2-ethoxyethoxy)ethan-1-ol SMILES: CCOCCOCCO
2-(2-Ethoxyethoxy)ethanol, BAKER™, J.T. Baker™
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: diethylene glycol monoethyl ether,2-2-ethoxyethoxy ethanol,carbitol,transcutol,ethoxy diglycol,dioxitol,ethyl carbitol,ethyl digol,carbitol solvent,ethanol, 2-2-ethoxyethoxy PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
HYDRANAL™ NEXTGEN, Solstice
Titrating agent for volumetric, one-component Karl Fischer titrations (Methanol-free)
2(2-Ethoxyethoxy)ethanol, 98+%
CAS: 111-90-0 MDL Number: MFCD00002872 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: diethylene glycol monoethyl ether,2-2-ethoxyethoxy ethanol,carbitol,transcutol,ethoxy diglycol,dioxitol,ethyl carbitol,ethyl digol,carbitol solvent,ethanol, 2-2-ethoxyethoxy PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
Diethylene Glycol Monoethyl Ether 99.0+%, TCI America™
Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 MDL Number: MFCD00002872 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: Carbitol, 2-(2-Ethoxyethoxy)ethanol PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
HydroSpec™ Composit T2, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
Diethylene Glycol Monoethyl Ether (Laboratory), Fisher Chemical
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 MDL Number: MFCD00002872 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: diethylene glycol monoethyl ether,2-2-ethoxyethoxy ethanol,carbitol,transcutol,ethoxy diglycol,dioxitol,ethyl carbitol,ethyl digol,carbitol solvent,ethanol, 2-2-ethoxyethoxy PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Solstice
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 MDL Number: MFCD00002872 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N Synonym: diethylene glycol monoethyl ether,2-2-ethoxyethoxy ethanol,carbitol,transcutol,ethoxy diglycol,dioxitol,ethyl carbitol,ethyl digol,carbitol solvent,ethanol, 2-2-ethoxyethoxy PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
Di(ethylene glycol)ethyl ether, SPEX CertiPrep™
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO
Carbitol, SPEX CertiPrep™
CAS: 111-90-0 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 InChI Key: XXJWXESWEXIICW-UHFFFAOYSA-N PubChem CID: 8146 ChEBI: CHEBI:40572 IUPAC Name: 2-(2-ethoxyethoxy)ethanol SMILES: CCOCCOCCO